missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUZ12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584345
This item is not returnable.
View return policy
Description
Immunogen sequence: NKPGSVKPTQ TIAVKESLTT DLQTRKEKDT PNENRQKLRI FYQFLYNNNT RQQTEARDDL HCPWCTLNCR KLYSLLKHLK LCHSRFIFNY VYHPKGARID VSINECYDGS YAGNPQDIHR QPGFAFSRNG PVKRTPIT Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 90%.
SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Specifications
| SUZ12 | |
| Polyclonal | |
| Unconjugated | |
| SUZ12 | |
| 2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15022 | |
| SUZ12 | |
| Recombinant protein corresponding to Human SUZ12. Recombinant protein control fragment (Product #RP-102304). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion