missing translation for 'onlineSavingsMsg'
Learn More

Suvorexant, Mikromol™

Product Code. 30050430
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050430 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050430 Supplier Mikromol Supplier No. MM3792.000250
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Suvorexant
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Suvorexant
CAS 1030377-33-3
Analyte or Component Names Suvorexant
Shipping Condition Room Temperature
Molecular Formula C23 H23 Cl N6 O2
Certifications/Compliance ISO 17025
Synonym Methanone, [(7R)-4-(5-chloro-2-benzoxazolyl)hexahydro-7-methyl-1H-1,4-diazepin-1-yl][5-methyl-2-(2H-1,2,3-triazol-2-yl)phenyl]-, [(7R)-4-(5-Chloro-2-benzoxazolyl)hexahydro-7-methyl-1H-1,4-diazepin-1-yl][5-methyl-2-(2H-1,2,3-triazol-2-yl)phenyl]methanone, 5-Chloro-2-[(5R)-5-methyl-4-[5-methyl-2-(2H-1,2,3-triazol-2-yl)benzoyl]-1,4-diazepan-1-yl]-1,3-benzoxazole, Belsomra, MK 4305, [(R)-4-(5-Chloro-benzooxazol-2-yl)-7-methyl-[1,4]diazepan-1-yl]-(5-methyl-2-[1,2,3]triazol-2-yl-phenyl)-methanone
InChI Formula InChI=1S/C23H23ClN6O2/c1-15-3-5-20(30-25-8-9-26-30)18(13-15)22(31)29-12-11-28(10-7-16(29)2)23-27-19-14-17(24)4-6-21(19)32-23/h3-6,8-9,13-14,16H,7,10-12H2,1-2H3/t16-/m1/s1
SMILES C[C@@H]1CCN(CCN1C(=O)c2cc(C)ccc2n3nccn3)c4oc5ccc(Cl)cc5n4
IUPAC Name [(7R)-4-(5-chloro-1,3-benzoxazol-2-yl)-7-methyl-1,4-diazepan-1-yl]-[5-methyl-2-(triazol-2-yl)phenyl]methanone
Molecular Weight (g/mol) 450.92
Quantity 250 mg
Formula Weight 450.1571 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.