missing translation for 'onlineSavingsMsg'
Learn More

Suvecaltamide, MedChemExpress

Product Code. 16479806
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479806 50 mg 50mg
16455987 5 mg 5mg
16423268 10 mg 10mg
16464526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479806 Supplier Medchem Express Supplier No. HY101096
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Suvecaltamide (MK-8998; compound 33) is a potent and selective inhibitor of the T-type calcium channel.

Suvecaltamide (MK-8998

Specifications

Chemical Name or Material Suvecaltamide
CAS 953778-58-0
Color White
Purity Grade Notes Research
Molecular Formula C20H23F3N2O2
Quantity 50 mg
Synonym MK-8998
Solubility Information DMSO : 100 mg/mL (262.88 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C1=NC=C(OCC(F)(F)F)C=C1)C)CC2=CC=C(C(C)C)C=C2
Molecular Weight (g/mol) 380.4
Formula Weight 380.4
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.