missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUV420H2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115484
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KMT5C/SUV4-20h2.
SUV420H2 is a histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. The protein mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. SUV420H1 is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2).
Specifications
| SUV420H2 | |
| Polyclonal | |
| Unconjugated | |
| KMT5C | |
| BC024816; Histone-lysine N-methyltransferase KMT5C; histone-lysine N-methyltransferase SUV420H2; KMT5C; lysine (K)-specific methyltransferase 5C; lysine methyltransferase 5C; lysine N-methyltransferase 5C; Lysine-specific methyltransferase 5C; PP7130; Su(var)4-20 homolog 2; suppressor of variegation 4-20 homolog 2; suppressor of variegation 4-20 homolog 2 (Drosophila); SUV420H2; suv4-20h2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232811, 308345, 84787 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P0C2N6, Q6Q783, Q86Y97 | |
| KMT5C | |
| A synthesized peptide derived from human KMT5C(Accession Q86Y97), corresponding to amino acid residues H92-I142. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido