Learn More
SUV420H2 Antibody (RAB-C409) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20151-0.025mg
 View more versions of this product
Product Code. 18684714
  • £190.00 / 0.02mg
Description
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SUV420H2
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
84787
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry
RAB-C409
Unconjugated
EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2
This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions.
0.025 mg
Primary
Human
Purified