missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420H2 Antibody (RAB-C409) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20151-0.2mg
This item is not returnable.
View return policy
Description
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Specifications
| SUV420H2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84787 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry | |
| RAB-C409 | |
| Unconjugated | |
| EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions. | |
| 0.2 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction