missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUV420H1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566611
This item is not returnable.
View return policy
Description
Immunogen sequence: DQGEHSGTVGV PVSYTDCAPS PVGCSVVTSD SFKTKDSFRT AKSKKKRRIT RYDAQLILEN NSGIPKLTLR Highest antigen sequence identity to the following orthologs - mouse 86%, rat 87%.
Histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. Mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. KMT5B is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2). Plays a role in myogenesis by regulating the expression of target genes, such as EID3.
Specifications
| SUV420H1 | |
| Polyclonal | |
| Unconjugated | |
| KMT5B | |
| [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; AA117471; C630029K18Rik; CGI85; CGI-85; histone-lysine N-methyltransferase KMT5B; histone-lysine N-methyltransferase SUV420H1; Kmt5b; lysine (K)-specific methyltransferase 5B; lysine methyltransferase 5B; lysine N-methyltransferase 5B; Lysine-specific methyltransferase 5B; MGC118906; MGC118909; MGC21161; MGC703; si:dkey-12e7.2; Su(var)4-20 homolog 1; suppressor of variegation 4-20 homolog 1; suppressor of variegation 4-20 homolog 1 (Drosophila); suppressor of variegation 4-20 protein-like 1; SUV420; SUV420H1; suv4-20h1; Unknown (protein for MGC:137299); wu:fb97g06; wu:fi57g03; zgc:103527 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4FZB7 | |
| KMT5B | |
| Recombinant Human SUV420H1. Recombinant protein control fragment (Product #RP-107271). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction