missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV3L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | SUV3L1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:4000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231930
|
Novus Biologicals
NBP3-38631-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231391
|
Novus Biologicals
NBP3-38631-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV3L1 Polyclonal antibody specifically detects SUV3L1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| SUV3L1 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ATP-dependent RNA helicase SUPV3L1, mitochondrial, EC 3.6.1, EC 3.6.4.13, suppressor of var1 (S.cerevisiae) 3-like 1, Suppressor of var1 3-like protein 1, suppressor of var1, 3-like 1 (S. cerevisiae), SUV3-like protein 1, SUV3suppressor of var1, 3-like 1(SUV3) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 577-786 of human SUV3L1 (NP_003162.2).,, Sequence:, APINKKQPFVCSSLLQFARQYSRNEPLTFAWLRRYIKWPLLPPKNIKDLMDLEAVHDVLDLYLWLSYRFMDMFPDASLIRDLQKELDGIIQDGVHNITKLIKMSETHKLLNLEGFPSGSQSRLSGTLKSQARRTRGTKALGSKATEPPSPDAGELSLASRLVQQGLLTPDMLKQLEKEWMTQQTEHNKEKTESGTHPKGTRRKKKEPDSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:4000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6832 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title