missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV39H2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38202-20ul
This item is not returnable.
View return policy
Description
SUV39H2 Polyclonal antibody specifically detects SUV39H2 in Human,Mouse,Rat samples. It is validated for ChIP assay,ELISA,Western Blot
Specifications
| SUV39H2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 5ug antibody for 10ug-15ug of Chromatin, ELISA | |
| FLJ23414Histone H3-K9 methyltransferase 2, H3-K9-HMTase 2, histone-lysine N-methyltransferase SUV39H2, KMT1BEC 2.1.1.43, Lysine N-methyltransferase 1B, Su(var)3-9 homolog 2, Suppressor of variegation 3-9 homolog 2, suppressor of variegation 3-9 homolog 2 (Drosophila) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 141-350 of human SUV39H2 (NP_001180354.1).,, Sequence:, CCPAEAGVLLAYNKNQQIKIPPGTPIYECNSRCQCGPDCPNRIVQKGTQYSLCIFRTSNGRGWGVKTLVKIKRMSFVMEYVGEVITSEEAERRGQFYDNKGITYLFDLDYESDEFTVDAARYGNVSHFVNHSCDPNLQVFNVFIDNLDTRLPRIALFSTRTINAGEELTFDYQMKGSGDISSDSIDHSPAKKRVRTVCKCGAVTCRGYLN | |
| 20 μL | |
| Cell Cycle and Replication | |
| 79723 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ChIP Assay, ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction