missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV39H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10574-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.Specifications
| SUV39H1 | |
| Polyclonal | |
| Unconjugated | |
| SUV39H1 | |
| H3 K9 HMTase 1, KMT1A, Lysine N methyltransferase 1A, MG44, Su(var)3 9 homolog 1, SUV39H, SUV39H1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20937, 302553, 6839 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43463, O54864 | |
| SUV39H1 | |
| SUV39H1 Fusion Protein Ag0854 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction