Learn More
Invitrogen™ Sutimlimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558457
 View more versions of this product
Product Code. 30284044
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sutimlimab is an anti-C1s antibody. Studies have shown that it can significantly increase hemoglobin levels in patients with primary cold agglutinin disease. Cold agglutinin disease (CAD) is a type of acquired autoimmune hemolytic anemia (AIHA). In cold environments, cold parts of the body will have clinical symptoms related to red blood cell aggregation (for example, the Internet Plaque or cyanosis) and hemolytic anemia.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sutimlimab Humanized
Recombinant Monoclonal
Unconjugated
BIVV009; IPN-009; TNT009
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized