missing translation for 'onlineSavingsMsg'
Learn More

Sutezolid, TRC

Product Code. 30419977
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30419977 5 mg -
30408820 25 mg -
30422060 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30419977

Brand: Toronto Research Chemicals TRCS8800035MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168828-58-8
Formula Weight 353.1209
InChI Formula InChI=1S/C16H20FN3O3S/c1-11(21)18-9-13-10-20(16(22)23-13)12-2-3-15(14(17)8-12)19-4-6-24-7-5-19/h2-3,8,13H,4-7,9-10H2,1H3,(H,18,21)/t13-/m0/s1
IUPAC Name N-[[(5S)-3-(3-fluoro-4-thiomorpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
Molecular Formula C16H20FN3O3S
Molecular Weight (g/mol) 353.41
SMILES CC(NC[C@H]1CN(C(O1)=O)C2=CC(F)=C(N3CCSCC3)C=C2)=O
Synonym N-[[(5S)-3-[3-Fluoro-4-(4-thiomorpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide, PNU 100480, U 100480
Chemical Name or Material Sutezolid

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.