missing translation for 'onlineSavingsMsg'
Learn More

Sutezolid, MedChemExpress

Product Code. 16452906
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452906

Brand: Medchem Express HY10392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sutezolid (PNU-100480), an orally active oxazolidinone antimicrobial agent, acts by inhibiting bacterial protein synthesis. Sutezolid has potent activity against mycobacteria, and is used for the research of drug-resistant tuberculosis.

Sutezolid (PNU-100480), an orally active oxazolidinone antimicrobial agent, acts by inhibiting bacterial protein synthesis. Sutezolid has potent activity against mycobacteria, and is used for the research of drug-resistant tuberculosis.

Specifications

Chemical Name or Material Sutezolid
Physical Form Powder
CAS 168828-58-8
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H20FN3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym PNU-100480 U-100480 PF-02341272
Solubility Information DMSO : 50 mg/mL (141.48 mM; Need ultrasonic)
SMILES O=C(O[C@H]1CNC(C)=O)N(C1)C2=CC(F)=C(N3CCSCC3)C=C2
Molecular Weight (g/mol) 353.41
Formula Weight 353.41
Percent Purity 98.87%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.