missing translation for 'onlineSavingsMsg'
Learn More

Sutezolid, MedChemExpress

Product Code. 16493908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493908 10 mg 10mg
16452906 25 mg 25mg
16414946 100 mg 100mg
16410277 50 mg 50mg
16406637 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493908 Supplier Medchem Express Supplier No. HY10392
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sutezolid (PNU-100480), an orally active oxazolidinone antimicrobial agent, acts by inhibiting bacterial protein synthesis. Sutezolid has potent activity against mycobacteria, and is used for the research of drug-resistant tuberculosis.

Sutezolid (PNU-100480), an orally active oxazolidinone antimicrobial agent, acts by inhibiting bacterial protein synthesis. Sutezolid has potent activity against mycobacteria, and is used for the research of drug-resistant tuberculosis.

Specifications

Chemical Name or Material Sutezolid
Physical Form Powder
CAS 168828-58-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H20FN3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym PNU-100480 U-100480 PF-02341272
Solubility Information DMSO : 50 mg/mL (141.48 mM; Need ultrasonic)
SMILES O=C(O[C@H]1CNC(C)=O)N(C1)C2=CC(F)=C(N3CCSCC3)C=C2
Molecular Weight (g/mol) 353.41
Formula Weight 353.41
Percent Purity 98.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.