Learn More
Poulten Selfe™ Suspended-Level Viscometers
Product Code. 13141053
Click to view available options
Size:
1
2
2A
3
3A
4
4A
5
Unit Size:
Each
Product Code. 13141053
Brand: Poulten Selfe™ 1625/09
Only null left
Add to basket
Add to basket
Suitable for transparent liquids. With UKAS certificate of calibration.
  • Type BS/IP/SL, IP71.Suitable for transparent liquids. Nominal overall length 330mm. Approximate sample size 22mL ­to 40mL. Available without certificate of calibration, please enquire.
  • With UKAS certificate of calibration.
Specifications
Product Type Suspended-Level Viscometer
Viscosity Range 20,000 to 1,000,000 cSt
Certifications/Compliance UKAS
Size 5
Volume (Metric) 22 to 40 mL