missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Antibody (W5C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09108
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| SUSD2 | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry 1-4 ug/ml, Immunocytochemistry/Immunofluorescence | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| WERI-RB-1 retinoblastoma cells | |
| 0.1 mg | |
| Cancer | |
| 56241 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence | |
| W5C5 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction