missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin [p Thr34] Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB500-236CL1
This item is not returnable.
View return policy
Description
Survivin Polyclonal antibody specifically detects Survivin in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Survivin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| This Survivin [p Thr34] Antibody was developed against a synthetic peptide with a phosphorylated Threonine (amino acid 34) corresponding to human Survivin. [UniProt# O15392] | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction