missing translation for 'onlineSavingsMsg'
Learn More

Survivin Monoclonal Antibody (STLALYV), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£171.00

Specifications

Antigen Survivin
Clone STLALYV
Concentration 0.5mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15587586
View Documents
Invitrogen
14-9176-80
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Regulated inhibition of programmed cell death (apoptosis) preserves normal homeostasis and tissue and organ morphogenesis. Aberrations in this process contribute to human diseases and cancer by abnormally prolonging cell viability. Recently, several apoptosis inhibitors related to the baculovirus iap gene have been found in various species, including human. IAP proteins contain one/three Cys/His baculovirus IAP repeats plus a c-terminus RING finger and are thought to block an evolutionary conserved step in apoptosis. Survivin encodes a structurally unique inhibitor of apoptosis (IAP). Survivin expression is turned off during fetal development and is not found in non-neoplastic adult human tissues. Survivin becomes abundantly re-expressed in transformed cells and in all of the most common cancers of lung, colon, pancreas, breast and prostate in vivo. Survivin appears to be situated at the crossroads of cell death and cell division, governing a checkpoint involved in cytokinesis while also suppressing apoptosis. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury.

Survivin (IAP4) is the smallest member of the Inhibitors of Apoptosis Protein (IAP) gene family. The IAPs are involved in multiple cell functions such as cell signaling, cell division, metabolism, and exhibits differential expression in nearly all human cancers, but not in most normal tissues. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but the Survivin gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of survivin’s gene expression. Survivin is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. Survivin may counteract a default induction of apoptosis in G2/M phase. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.
Specifications

Specifications

Survivin
0.5mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
API4, Apoptosis Inhibitor 4, Apoptosis inhibitor survivin, Baculoviral IAP repeat containing 5, BIRC 5, EPR 1, IAP4, SVV, TIAP
BIRC5
Primary
Store at 2-8°C.
BIRC5
STLALYV
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
O15392
332
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.