missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (CL12064), Novus Biologicals™
Mouse Monoclonal Antibody
£277.00 - £415.00
Specifications
| Antigen | Survivin |
|---|---|
| Clone | CL12064 |
| Dilution | Western Blot 1 μg/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639544
|
Novus Biologicals
NBP3-18538-100ul |
100 μg |
£415.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678434
|
Novus Biologicals
NBP3-18538-25ul |
25 μg |
£277.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Western BlotSpecifications
| Survivin | |
| Western Blot 1 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| This antibody was developed against a sythetic peptide for O15392. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12064 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| PBS, pH 7.2, 40% glycerol | |
| 332 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title