Learn More
Survivin Antibody (CL12064) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18538AF350
Product Code. 30290598
  • £582.00 / 0.10mL
Description
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Survivin
Monoclonal
Alexa Fluor 350
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
This antibody was developed using a synthetic peptide derived from O15392, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
332
Store at 4⪚C in the dark.
IgG1
Western Blot
CL12064
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified