missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
Specifications
| Antigen | Survivin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 800 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha |
| Host Species | Mouse |
| Immunogen | This Survivin Antibody (8E2) was developed against full length recombinant human Survivin [UniProt# O15392] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?