missing translation for 'onlineSavingsMsg'
Learn More

Survivin Antibody (32.1) - Azide and BSA Free, Novus Biologicals™

Product Code. 18384110 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384110 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384110 Supplier Novus Biologicals Supplier No. NBP280968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Survivin Monoclonal specifically detects Survivin in Human, Hamster samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Survivin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 32.1
Concentration 1.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry, ELISA, Immunohistochemistry : 20 μg/mL, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 20 μg/mL, CyTOF-ready
Formulation PBS
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Gene Symbols BIRC5
Host Species Mouse
Immunogen This Survivin Antibody (32.1) was developed against full length recombinant human Survivin [UniProt# O15392]
Molecular Weight of Antigen 16 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 332
Test Specificity Human and hamster.
Target Species Human, Hamster
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.