missing translation for 'onlineSavingsMsg'
Learn More

Polynuclear Aromatic Hydrocarbons Surrogate Standard, SPEX CertiPrep™

Product Code. 11470259 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11470259

Brand: SPEX Certiprep 8100S

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1-Fluoronaphthalene321-38-02000μg/mL
2-Fluorobiphenyl321-60-82000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 434-90-2
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Method 8100
Molecular Formula C12F10
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene
Solvent or Matrix MeCl
Molecular Weight (g/mol) 334.116
PubChem CID 67949
Quantity 1 mL
Chemical Name or Material PAH Surrogate Standard, SW-846 Method 8100
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • PAH Surrogate Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : dichloromethane

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.