missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Surfactant Protein A Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-12928
This item is not returnable.
View return policy
Description
Surfactant Protein A Polyclonal specifically detects Surfactant Protein A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Surfactant Protein A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| SFTPA1 | |
| Peptide with sequence C-HLDEELQATLHDFR corresponding to internal region according to NP_005402.3, NP_001087239.2, NP_001158117.1, NP_001158118.1, NP_001092138.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize all the reported isoforms of A1 (NP_005402.3; NP_001087239.2; NP_001158117.1; NP_001158118.1) and A2 ( NP_001092138.1), Reported variants represent identical protein: NP_001158116.1, NP_005402.3, NP_001158119.1 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.01 ug/ml, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 653509 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction