Learn More
Neogen Surface Protein Plus
Product Code. 17283734
Encompass
Change view
Click to view available options
No. per Case:
50 /Cs.
Unit Size:
50 tests
Product Code. No. per Case unitSize
17283734 50 /Cs. 50 tests
1 options
Product Code. 17283734 Brand Neogen Brand No. 7000086286
Simple, quick, and reliable method of checking for the removal of protein residues after cleaning and quickly identifies contamination.
  • Helps to reduce the risk of a food safety incident.
  • Detects as little as 50μg of protein on surfaces and in solution.
  • Tests for a the presence of proteins to verify effectiveness of cleaning.
Specifications
No. per Case 50 /Cs.
Product Type Test Swab
compliance-icons
Product Identifier
  • X50 Clean-Trace Surface Protein Plus test - PRO50
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : copper sulphate pentahydrate