Learn More
Miele Professional™ Cover
Filter E 319/3 for optimum protection against contamination from labels or glass splinters
Brand:  Miele Professional™ 6477070
Additional Details : Weight : 0.50000kg
Product Code. 10673014
  • £357.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Surface filter for coarse waste
  • Prevents soiling by labelling laboratory glassware
  • Prevents contamination with splinters of broken glass
Specifications
Specifications
Stainless Steel
Stainless Steel
Washer-disinfectors, laboratory
500 mm
Cover
E 319/3 Insert 1/1
488 mm
1 Each