missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SURF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567356
This item is not returnable.
View return policy
Description
Immunogen sequence: KEKRQRVKGNL TPLTGRNYRQ LLERLQARQS RLDELRGQDE GKAQELEAKM KWTNLLYKAE GVKIRDDERL L Highest antigen sequence identity to the following orthologs - mouse 92%, rat 89%.
This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene demonstrates features of a housekeeping gene, being ubiquitously expressed, and the encoded protein has been localized to the nucleolus. The protein includes motifs found in both the mouse and fish orthologs, which suggests a putative function as a nucleolar-matrix protein with nucleic acid-binding properties, based on characteristics determined in mouse.This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene demonstrates features of a housekeeping gene, being ubiquitously expressed, and the encoded protein has been localized to the nucleolus. The protein includes motifs found in both the mouse and fish orthologs, which suggests a putative function as a nucleolar-matrix protein with nucleic acid-binding properties, based on characteristics determined in mouse.
Specifications
| SURF6 | |
| Polyclonal | |
| Unconjugated | |
| SURF6 | |
| D2Wsu129e; RRP14; SURF6; Surf-6; surfeit 6; surfeit gene 6; surfeit locus protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75683 | |
| SURF6 | |
| Recombinant Human SURF6. Recombinant protein control fragment (Product #RP-105590). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction