missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SURF5 Monoclonal antibody specifically detects SURF5 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | SURF5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PCRP-MED22-2A7 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human SURF5 protein (Uniprot: Q15528) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?