missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SURF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565711
This item is not returnable.
View return policy
Description
Immunogen sequence: GDGTDDFLTDK EDEKAKPPRE KATDESRRET TVYRGLVQKR GKKQLGSLKK KFKSHHRKPK SFSSCKQ Highest antigen sequence identity to the following orthologs - mouse 53%, rat 41%.
This gene shares a bidirectional promoter with surfeit 1 (SURF1; GeneID: 6834), which is located on the opposite strand. It encodes a conserved protein that is expressed in a variety of tissues.
Specifications
| SURF2 | |
| Polyclonal | |
| Unconjugated | |
| SURF2 | |
| SURF2; Surf-2; surfeit 2; surfeit gene 2; surfeit locus protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15527 | |
| SURF2 | |
| Recombinant Human SURF2. Recombinant protein control fragment (Product #RP-106382). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction