missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay

Product Code. 17839132 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Product Code. 17839132 Supplier Thermo Scientific™ Supplier No. A56839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly detect Staphylococcus aureus in food samples with the Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay.

Rapidly detect Staphylococcus aureus in food and environmental samples with the Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay.

The AOAC-RI validation covers dairy products including infant formula together with other low moisture products and cheeses.

The assay is validated by AOAC-RI and enables users to reliably detect Staphylococcus aureus from dairy products, including infant formula and other low moisture dairy products and cheeses, in less than 24 hours.

The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.

Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 24 hours
  • Maximum flexibility for operational efficiency
  • Run all assays simultaneously in the same run
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps
  • Detect multiple targets in the same run with universal PCR protocols across the range

Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results

The Thermo Scientific SureTect Staphylococcus aureus PCR Assay includes:

  • Lysis Reagent 1 Tubes
  • Lysis Reagent 2 
  • Proteinase K
  • Staphylococcus aureus PCR Tubes
  • Thermo Scientific 96-Well Pierceable Seals
  • PCR Caps

Enriched samples are pipetted into pre-filled lysis tubes, along with Proteinase K, before incubation, to lyse any bacterial cells present in the sample and release their DNA into the solution. The lysate is then loaded into the SureTect Staphylococcus aureus PCR tube to re-hydrate the pre-loaded PCR pellet which contains all the necessary components and reagents for PCR, including a probe, primers, and DNA template for the internal positive control (IPC).

The PCR tubes are then sealed, loaded into the Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR instrument using RapidFinder™ Analysis Software v3.0 or later or the Applied Biosystems 7500 Fast Food Safety 96-well Real-Time PCR instrument using the RapidFinder™ Express Software v2.0 or later. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required. The internal postive control confirms correct assay conditions have occurred.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Product Type SureTect S. aureus Assay
Target Organism Class Staphylococcus aureus
For Use With (Application) Food and Beverage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.