Learn More
Thermo Scientific™ SureTect™ Salmonella Species PCR Assay
Description
Detect Salmonella species in even the most challenging food types with this multiplex real-time assay that provides highly-specific detection of the invA gene.
Detect Salmonella species in food and environmental samples with the Thermo Scientific™ SureTect™ Salmonella Species PCR Assay which incorporates optimized primer and probe components for highly-specific detection of Salmonella species. The assay has been AOAC-RI and NF VALIDATION™ validated with a variety of matrices and a range of samples sizes and enrichments, following the AOAC or NF validated methods, to reduce the need for in-house validation prior to use. The internal amplification control confirms correct assay conditions have occurred and results can be relied on.
- Rapid actions: fast, optimized single enrichment step and automatic result interpretation
- Simple workflow: single enrichment protocols, pre-dispensed reagents, universal PCR protocols for detection of multiple targets in the same run.
- Intuitive, user-friendly software: reliable results with no subjective interpretation.
- Peace of mind: broad range of validated workflows according to NF validation, AFNOR and AOAC-RI certified, supported by our dedicated and experienced technical teams.
- Maximum flexibility: the ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
The SureTect Salmonella Species PCR Assay has been extensively tested on over 20 different matrices from food manufacturing environments including meat, dairy products,fish, fruit and vegetables, and production surfaces. Specific samples tested during independent external validation include food types most commonly associated with contamination by Salmonella species and also those food types known to be particularly challenging to PCR detection, for example, ground beef, raw poultry, eggs, non-fat dried milk and ready-to-eat meals.
Includes:
- SureTect Lysis Reagent 1 Tubes
- SureTect Lysis Tube Caps
- SureTect Proteinase K
- SureTect Salmonella PCR Tubes
- SureTect PCR Caps
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. Thermo Fisher Scientific has evaluated the SureTect PCR Assay workflows with industry leading Oxoid™ and Remel™ media formulations, as well as, ISO formulations where fit for purpose.
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Content And Storage | All the necessary reagents for DNA release and real-time PCR for 96 tests. Store in the dark at 2° to 8°C. |
| Description | SureTect Salmonella Assay |
| Detection Method | qPCR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| Reaction Speed | Fast |
| For Use With (Equipment) | Applied Biosystems 7500 Fast Real-Time PCR instrument and Applied Bisystems QuantStudio 5 Real Time PCR System |
| No. of Tests | 96 Tests/Kit |
| Product Type | qPCR Reagents |
| For Use With (Application) | Food and Beverage |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.