Learn More
Thermo Scientific™ SureTect™ E. coli O157:H7 PCR Assay
Description
Detect E. coli O157:H7 rapidly with the highly-specific, real-time Thermo Scientific™ SureTect™ E. coli O157:H7 PCR Assay. The assay incorporates an optimized liquid lysis step, providing a quick and simple method for preparing the sample for rapid detection on the Applied Biosystems™ 7500 Fast Real-Time PCR instruments or Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR System. The assay has been validated with a variety of matrices, samples sizes, and enrichment volumes, following the AOAC, USDA MLG or NF validated methods, to reduce the need for in-house validation prior to use.
- Rapid actions: fast, optimized single enrichment step and automatic result interpretation.
- Simple workflow: single enrichment protocols, pre-dispensed reagents, universal PCR protocols for detection of multiple targets in the same run.
- Intuitive, user-friendly software: reliable results, with no subjective interpretation.
- Peace of mind: broad range of validated workflows according to AFNOR and AOAC-RI, supported by our dedicated and experienced technical teams
- Maximum flexibility: the ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
The SureTect E. coli O157:H7 PCR Assay has been extensively tested on a range of food matrices from food manufacturing environments, including samples of raw ground beef, raw beef trim, fruit juice and produce.
Includes:
- SureTect Lysis Reagent 1 Tubes
- SureTect Lysis Tube Caps
- SureTect Proteinase K
- SureTect E. coli O157:H7 PCR Tubes
- SureTect PCR Caps
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. Thermo Fisher Scientific has evaluated the SureTect PCR Assays workflows with industry leading Oxoid™ and Remel™ media formulations, as well as, ISO formulations where fit for purpose.
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Description | SureTect E. coli O157:H7 PCR Assay |
| Detection Method | Primer-probe |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| Reaction Speed | Fast |
| For Use With (Equipment) | Applied Biosystems 7500 Fast Real-Time PCR instrument and Applied Bisystems QuantStudio 5 Real Time PCR System |
| No. of Tests | 96 Tests/Kit |
| Product Type | qPCR Reagents for Microbial Detection |
| For Use With (Application) | Real Time PCR (qPCR) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.