Learn More
Invitrogen™ SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, PVDF
Description
The 0.45-μm pore size makes these membranes ideal for protein transfer for proteins >10 kDa. The dimensions (8.6 x 14 cm) of these PVDF membrane and filter paper sandwiches are ideal for wet-transfer of Invitrogen midi gels using the SureLock Tandem Midi Blot Module. The 0.45-μm pore size membrane and filter paper sandwiches are conveniently supplied as pack of 20.
Features:
- Membrane: polyvinylidene difluoride (PVDF)
- Filter paper thickness: 0.8 mm
- Re-probe characteristics: yes
- Pre-activation: required with 100% alcohol (methanol, ethanol, or isopropanol)
- Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
- Durability: compatible with most organic solvents, acids, and mild bases
- Dimensions: 8.6 x 14 cm
Specifications
Specifications
| Quantity | 20 Pre-cut Membranes and Filters, 0.45 μm, PVDF Sandwich |
| Format | Sandwich |
| Material | PVDF |
| Pore Size | 0.45 μm |
| Content And Storage | Contains 20 pre-cut membrane and filter sandwiches (0.45 μm, PVDF) |
| Dimensions (LxW) | Midi, 14 cm x 8.6 cm |
| Product Line | SureLock |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.