Learn More
Invitrogen™ SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose
Invitrogen SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose, are high-quality membranes ideal for blotting proteins and nucleic acids.
Brand: Invitrogen™ STM2008
Additional Details : Weight : 2.26000kg
Description
The 0.45-μm pore size is ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp). The dimensions (8.6 x 14 cm) of these nitrocellulose membrane and filter paper sandwiches are ideal for wet-transfer of Invitrogen midi gels using the SureLock Tandem Midi Blot Module. The 0.2 μm pore size membrane and filter paper sandwiches are conveniently supplied as pack of 20.
Features:
- Membrane: 100% pure nitrocellulose
- Filter paper thickness: 0.8 mm
- Re-probe characteristics: yes
- Binding interactions: hydrophobic and electrostatic
- Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
- Dimensions: 8.6 x 14 cm
Specifications
20 Pre-cut Membranes and Filters, 0.45 μm, Nitrocellulose Sandwich | |
Nitrocellulose | |
Contains 20 pre-cut membrane and filter sandwiches (0.45 μm, nitrocellulose) | |
SureLock |
Sandwich | |
0.45 μm | |
Midi, 14 cm x 8.6 cm |