missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SureCast™ Stacking Buffer
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
Pack of 2
Pack of 5
Description
- Pour polyacrylamide gels easily and confidently
- For use with the SureCast vertical gel system
- SureCast gels are designed for use in the Mini Gel Tank
Recommended Storage
Room Temperature
Specifications
Specifications
| Content And Storage | 5 x 500 mL (2.5 L total) SureCast Stacking Buffer packs. Store at room temperature. |
| Product Type | Stacking Buffer (2.5 L) |
| Product Line | SureCast |
| Quantity | 5 x 500 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction