missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SureCast™ Multi-Use Tool, 12-well
Description
- Pour polyacrylamide gels easily and confidently
- For use with the SureCast vertical gel system
- SureCast gels are designed for use in the Mini Gel Tank
Recommended Storage
Room Temperature
Warranty
Up to 12 months
Specifications
Specifications
| For Use With (Equipment) | SureCast Vertical Gel System |
| Gel Size | Mini |
| Product Line | SureCast |
| Type | 12-well Multi-Use Tool |
| Quantity | 1 unit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?