missing translation for 'onlineSavingsMsg'
Learn More

Suramin (Sodium Salt), TRC

Product Code. 30011765
Change view
Click to view available options
Quantity:
5 mg
50 mg
500 mg
Unit Size:
500mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011765 500 mg 500mg
30137096 50 mg 50mg
30008472 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011765 Supplier Toronto Research Chemicals Supplier No. TRCS860000500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-46-4
Molecular Formula C51 H34 N6 O23 S6 . 6 Na
Molecular Weight (g/mol) 1429.17
Synonym Antrypol, Bayer 205, Germanin, Moranyl, Naganine, Suramine sodium, 8,8'-(Carbonylbis(imino-3,1-phenylenecarbonylimino(4-methyl-3,1-phenylene)carbonylimino))bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
IUPAC Name hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].Cc1ccc(cc1NC(=O)c2cccc(NC(=O)Nc3cccc(c3)C(=O)Nc4cc(ccc4C)C(=O)Nc5ccc(c6cc(cc(c56)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])c2)C(=O)Nc7ccc(c8cc(cc(c78)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C51H40N6O23S6.6Na/c1-25-9-11-29(49(60)54-37-13-15-41(83(69,70)71)35-21-33(81(63,64)65)23-43(45(35)37)85(75,76)77)19-39(25)56-47(58)27-5-3-7-31(17-27)52-51(62)53-32-8-4-6-28(18-32)48(59)57-40-20-30(12-10-26(40)2)50(61)55-38-14-16-42(84(72,73)74)36-22-34(82(66,67)68)24-44(46(36)38)86(78,79)80;;;;;;/h3-24H,1-2H3,(H,54,60)(H,55,61)(H,56,58)(H,57,59)(H2,52,53,62)(H,63,64,65)(H,66,67,68)(H,69,70,71)(H,72,73,74)(H,75,76,77)(H,78,79,80);;;;;;/q;6*+1/p-6
Formula Weight 1427.9386
Chemical Name or Material Suramin Hexasodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.