missing translation for 'onlineSavingsMsg'
Learn More

Suramin sodium salt, MedChemExpress

Product Code. 16414507
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16414507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414507 Supplier Medchem Express Supplier No. HYB0879A50mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Suramin sodium salt (Suramin hexasodium salt) is a reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor.

Suramin sodium salt is a potent inhibitor of sirtuins: SirT1 (IC50=297 nM), SirT2 (IC50=1.15 μM), and SirT5 (IC50=22 μM). Suramin sodium salt is a competitive inhibitor of reverse transcriptase (DNA topoisomerase II: IC50=5 μM). Suramin sodium salt is a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor. Suramin sodium salt efficiently inhibits IP5K and is an antiparasitic, anti-neoplastic and anti-angiogenic agent.

Specifications

Chemical Name or Material Suramin sodium salt
Physical Form Solid
CAS 129-46-4
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C51H34N6Na6O23S6
For Use With (Application) Cancer-programmed cell death
Synonym Suramin hexasodium salt
Solubility Information H2O : ≥ 200 mg/mL (139.94 mM) ∣DMSO : 83.33 mg/mL (58.31 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(NC2=CC(C(NC3=CC=C(S(=O)(O[Na])=O)C4=CC(S(=O)(O[Na])=O)=CC(S(=O)(O[Na])=O)=C34)=O)=CC=C2C)=O)=CC=C1)NC5=CC(C(NC6=CC(C(NC7=CC=C(S(=O)(O[Na])=O)C8=CC(S(=O)(O[Na])=O)=CC(S(=O)(O[Na])=O)=C78)=O)=CC=C6C)=O)=CC=C5
Molecular Weight (g/mol) 1429.17
Formula Weight 1429.17
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.