missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUR1 Monoclonal Antibody (N289/16), APC

Description
1 μg/mL of MA5-45558 was sufficient for detection of SUR1 in 20 μg of mouse brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. Does not cross-react with SUR2B. This antibody was formerly sold as clone S289-16.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a modulator of ATP-sensitive potassium channels and insulin release. Mutations and deficiencies in this protein have been observed in patients with hyperinsulinemic hypoglycemia of infancy, an autosomal recessive disorder of unregulated and high insulin secretion. Mutations have also been associated with non-insulin-dependent diabetes mellitus type II, an autosomal dominant disease of defective insulin secretion. Alternative splicing of this gene has been observed; however, the transcript variants have not been fully described.
Specifications
Specifications
| Antigen | SUR1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N289/16 |
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Formulation | 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 |
| Gene | ABCC8 |
| Gene Accession No. | Q09428, Q09429 |
| Gene Alias | ABC36; ABCC8; AM60008PU-N; ATP binding cassette subfamily C member 8; ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, subfamily C (CFTR/MRP), member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; D930031B21Rik; HHF1; HI; HRINS; LOW QUALITY PROTEIN: ATP-binding cassette sub-family C member 8; MRP8; PH SUR1; PHHI; sulfonylurea receptor; sulfonylurea receptor (hyperinsulinemia); sulfonylurea receptor 1; sulfonylurea receptor subunit 1; sulphonylurea receptor 1; SUR; SUR1; SUR1 protein; SUR1delta2; TNDM2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?