missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUR1 Antibody (S289-16), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59320V
This item is not returnable.
View return policy
Description
SUR1 Monoclonal specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SUR1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ABCC8 | |
| Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 | |
| 100 μL | |
| Primary | |
| Detects 160kDa. Does not cross-react with SUR2B. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S289-16 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
| Mouse | |
| Protein G purified | |
| ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain | |
| 6833 | |
| Human, Mouse, Rat, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction