missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUPT7L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566610
This item is not returnable.
View return policy
Description
Immunogen sequence: PTMLDIPSEPC SLTIHTIQLI QHNRRLRNLI ATAQAQNQQQ TEGVKTEESE PLPSCPGSPP LPDDLLPLDC KNPNAPFQIR HSD Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.
SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.
Specifications
| SUPT7L | |
| Polyclonal | |
| Unconjugated | |
| SUPT7L | |
| 2610524B01Rik; 6030455L14Rik; Adenocarcinoma antigen ART1; KIAA0764; RGD1562206; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPT7L; SPT7-like STAGA complex gamma subunit; SPTF-associated factor 65 gamma; STAF65; STAF65(gamma); STAF65G; STAF65gamma; STAGA complex 65 gamma subunit; STAGA complex 65 subunit gamma; STAGA complex 65 subunit gamma-like; suppressor of Ty 7 (S. cerevisiae)-like; suppressor of Ty 7-like; SUPT7H; SUPT7L; Unknown (protein for MGC:137593) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9913 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94864 | |
| SUPT7L | |
| Recombinant Human SUPT7L. Recombinant protein control fragment (Product #RP-105265). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction