missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SUPT5H Antibody (3B1), Novus Biologicals™

Product Code. 18347169 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347169 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347169 Supplier Novus Biologicals™ Supplier No. H00006829M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SUPT5H Monoclonal antibody specifically detects SUPT5H in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT5H
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 3B1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH24203
Gene Alias DRB sensitivity-inducing factor 160 kDa subunit, DRB sensitivity-inducing factor large subunit, DSIF large subunit, DSIF p160, FLJ34157, SPT5hSPT5, SPT5HTat-cotransactivator 1 protein, suppressor of Ty (S.cerevisiae) 5 homolog, suppressor of Ty 5 homolog (S. cerevisiae), Tat-CT1, Tat-CT1 protein, transcription elongation factor SPT5
Host Species Mouse
Immunogen SUPT5H (AAH24203, 981 a.a. ∽ 1087 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TTDIQVKVRDTYLDTQVVGQTGVIRSVTGGMCSVYLKDSEKVVSISSEHLEPITPTKNNKVKVILGEDREATGVLLSIDGEDGIVRMDLDEQLKILNLRFLGKLLEA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6829
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.