missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT16H Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20551-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to SUPT16H.
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT , interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.Specifications
| SUPT16H | |
| Polyclonal | |
| Unconjugated | |
| SUPT16H | |
| CDC68, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTP140, hSPT16, SPT16/CDC68, SUPT16H | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11198, 114741 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q920B9, Q9Y5B9 | |
| SUPT16H | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction