missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT16H Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68243-1-IG-20UL
This item is not returnable.
View return policy
Description
The SUPT16H antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SUPT16H. This antibody recognizes human, mouse, rat antigen. The SUPT16H antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| SUPT16H | |
| Monoclonal | |
| 1000 μg/mL | |
| CDC68; FACTP140; FLJ10857; FLJ14010; FLJ34357; SPT16/CDC68 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 135 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1E4C4 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 11198 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction