missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT16H, Mouse anti-Human, Clone: 1D12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011198-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: PGEQTVPALNLQNAFRIIKEVQKRYKTREAEEKEKEGIVKQDSLVINLNRSNPKLKDLYIRPNIAQKRMQGSLEAHVNGFRFTSVRGDKVDILYNNIKHALFQPCDGESpecifications
| SUPT16H | |
| Monoclonal | |
| Unconjugated | |
| suppressor of Ty 16 homolog (S. cerevisiae) | |
| CDC68/FACT/FACTP140/FLJ10857/FLJ14010/FLJ34357/SPT16/CDC68 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1D12 | |
| In 1x PBS, pH 7.4 | |
| NM_007192 | |
| SUPT16H | |
| SUPT16H (NP_009123, 608 a.a. ∼ 715 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 11198 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction