missing translation for 'onlineSavingsMsg'
Learn More

SUPT16H Antibody (OTI8A1) [DyLight 650], Novus Biologicals™

Product Code. 30688332 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688332 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688332 Supplier Novus Biologicals Supplier No. NBP345025C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SUPT16H Monoclonal antibody specifically detects SUPT16H in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT16H
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI8A1
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 187-282 of human SUPT16H (NP_009123) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.