missing translation for 'onlineSavingsMsg'
Learn More

SUPT16H Antibody (OTI3B8) [DyLight 680], Novus Biologicals™

Product Code. 30688343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688343 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Produktkod. 30688343 Leverantör Novus Biologicals Leverantörsnummer NBP345016FR

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

SUPT16H Monoclonal antibody specifically detects SUPT16H in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen SUPT16H
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI3B8
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 187-282 of human SUPT16H (NP_009123) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.