missing translation for 'onlineSavingsMsg'
Learn More

SUPT16H Antibody (2D1), Novus Biologicals™

Product Code. 18370979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370979 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370979 Supplier Novus Biologicals Supplier No. H00011198M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SUPT16H Monoclonal antibody specifically detects SUPT16H in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT16H
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2D1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_009123
Gene Alias CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae)
Host Species Mouse
Immunogen SUPT16H (NP_009123, 608 a.a. ~ 715 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PGEQTVPALNLQNAFRIIKEVQKRYKTREAEEKEKEGIVKQDSLVINLNRSNPKLKDLYIRPNIAQKRMQGSLEAHVNGFRFTSVRGDKVDILYNNIKHALFQPCDGE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.