missing translation for 'onlineSavingsMsg'
Learn More

Microflex™ High Risk Disposable Nitrile Gloves

Product Code. 15523705
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50

Product Code. 15523705

Brand: Microflex 93853060

Only null left
Add to basket
Edge
Add to basket

Offer up to 3 times the puncture resistance of latex or vinyl disposable gloves. Microflex™ High Risk Disposable Nitrile Gloves also provide chemical and liquid protection.

  • Extended cuff provides additional protection for the wrist and arm for hazardous environments.
  • Advanced nitrile formulation provides the lowest acceptable pinhole rates (AQL 0.65)
  • Non-stick properties are built into the glove formulation to create a resistance to tape or adhesives, and to assist workers with improved efficiency and speed.
  • Textured fingers ensure strong, efficient grip and excellent worker protection.
  • Constructed without natural rubber latex materials or powder to help protect from allergies, skin irritation and dryness.
  • Approved for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, ISO 11193-1, EN420, 89/686/EEC, EN374-2: 2003, EN374-3: 2003, MDD 93/42/EEC, Class 1, EN455-1, EN455-2 and EN455-3, ASTM F1671
Applications Industrial
Color Blue
Disposable Single-use
Grade Medical
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Chemical-resistant Gloves
Size X-Small
Sterility Non-sterile
Thickness (Metric) 0.14 mm
Finger Style Textured
Glove Cuff Style Extended, Beaded
Model 93-853
Packaging Type Dispenser Box
Quantity 50Pack
Tensile Strength 14 to 30 MPa
Length (Metric) 295 mm
Width (Metric) 80 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.