missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suppressor of Ty 4 homolog 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93471-0.02ml
This item is not returnable.
View return policy
Description
Suppressor of Ty 4 homolog 1 Polyclonal antibody specifically detects Suppressor of Ty 4 homolog 1 in Human samples. It is validated for Western Blot
Specifications
| Suppressor of Ty 4 homolog 1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DRB sensitivity-inducing factor 14 kDa subunit, DRB sensitivity-inducing factor small subunit, DSIF p14, DSIF small subunit, SPT4, SPT4HhSPT4, suppressor of Ty (S.cerevisiae) 4 homolog 1, suppressor of Ty 4 homolog 1 (S. cerevisiae), SUPT4H, transcription elongation factor SPT4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human SUPT4H1 (NP_003159.1). MALETVPKDLRHLRACLLCSLVKTIDQFEYDGCDNCDAYLQMKGNREMVYDCTSSSFDGIIAMMSPEDSWVSKWQRVSNFKPGVYAVSVTGRLPQGIVRELKSRGVAYKSRDTAIKT | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6827 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction